Learn More
Medchemexpress LLC N-[(5-{[(4-methylphenyl)sulfonyl]amino}-3-(trifluoroacetyl)-1H-indol-1-yl)acetyl]-l-leucine | 1623002-61-8 | 98.2% | 553.55 | C25H26F3N3O6S | 1MG

Supplier: Medchemexpress LLC HY124361MG
NTRC-824 is a selective, non-peptide neurotensin receptor type 2 (NTS2) antagonist provided as a solid research compound. It is characterized by a molecular formula of C25H26F3N3O6S, a molecular weight of 553.55, and high purity, and is intended for in vitro pharmacology, receptor characterization, and screening applications.
- Selective NTS2 antagonist (IC50 38 nM).
- High purity ≥98.2% (HPLC).
- Molecular weight 553.55 and formula C25H26F3N3O6S.
- Off-white to light yellow solid appearance.
- Recommended storage: powder at -20°C; in solvent at -80°C for long-term.
- Supplied in a 1 mg quantity suitable for assay development and screening.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.