Learn More
Medchemexpress LLC N-[5-[(5-tert-butyl-1,3-oxazol-2-yl)methylsulfanyl]-1,3-thiazol-2-yl]piperidine-4-carboxamide | 345627-80-7 | MFCD09833875 | 99.9% | 380.53 | C17H24N4O2S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1000825MG
Encompass_Preferred
SNS-032 is a potent, ATP-competitive small-molecule inhibitor of cyclin-dependent kinases CDK9, CDK2, and CDK7. It is used in preclinical research to probe CDK-mediated control of transcription and the cell cycle and displays antitumor activity in experimental models. Reported IC50 values are 4 nM (CDK9), 38 nM (CDK2), and 62 nM (CDK7).
  • Potent inhibitor of CDK9, CDK2, and CDK7.
  • Low nanomolar potency supports sensitive biochemical assays.
  • ATP-competitive mechanism for target-specific profiling.
  • Suitable for in vitro and preclinical cellular studies.
  • Available as solid and ready-to-use DMSO solution formats.
  • High purity for reproducible experimental results.
Catalog No. 50-002-15435