Learn More
Medchemexpress LLC N-[5-[(5-tert-butyl-1,3-oxazol-2-yl)methylsulfanyl]-1,3-thiazol-2-yl]piperidine-4-carboxamide | 345627-80-7 | MFCD09833875 | 99.9% | 380.53 g/mol | C17H24N4O2S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1000850MG
Encompass_Preferred
SNS-032 is a small-molecule research compound that selectively inhibits cyclin-dependent kinases CDK2, CDK7, and CDK9 (IC50s: 38 nM, 62 nM, 4 nM, respectively). It is used in preclinical studies to probe cell-cycle regulation, transcriptional control, apoptosis induction, and angiogenesis inhibition.
  • Selective inhibition of CDK2, CDK7, and CDK9.
  • Potent transcriptional inhibitor reducing RNA polymerase II phosphorylation.
  • Demonstrated antitumor activity in preclinical cell models.
  • Reversible activity suitable for washout and time-course studies.
  • Available in small package sizes for laboratory research use.
Catalog No. 50-002-15367