Learn More
Medchemexpress LLC N-[5-(furan-2-yl)-1,3,4-oxadiazol-2-yl]-2-phenylquinoline-4-carboxamide | 851095-32-4 | 98.4% | 382.37 g/mol | C22H14N4O3 | 25 MG

Supplier: Medchemexpress LLC HY10369225MG
STX-0119 is a small-molecule inhibitor that selectively blocks STAT3 dimerization and suppresses STAT3-dependent transcription (reported IC50 ≈ 74 μM). It is provided for research use and is supplied as a solid for biochemical and cellular studies.
- Selective STAT3 dimerization inhibitor.
- Inhibits STAT3 transcription with IC50 ≈ 74 μM.
- Molecular formula C22H14N4O3; molecular weight 382.37 g/mol.
- High purity (98.4%) and white to light yellow solid appearance.
- Soluble in DMSO (13.33 mg/mL; may require warming and sonication).
- Store at 4°C protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.