Learn More
Medchemexpress LLC N-((6-methoxy-5-(trifluoromethyl)-1-naphthalenyl)thioxomethyl)-N-methylglycine | 82964-04-3 | ≥98.0% | 357.35 g/mol | C16H14F3NO3S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1650010MG
Encompass_Preferred
Tolrestat is a potent, orally active aldose reductase inhibitor (IC50 = 35 nM) supplied as a purified research standard for biochemical and pharmacological studies. The compound has formula C16H14F3NO3S, molecular weight 357.35 g/mol, and CAS 82964-04-3. Solubility: DMSO 50 mg/mL; aqueous formulations ≥ 2.5 mg/mL with appropriate solubilizers. Storage: powder at -20°C (up to 3 years); solutions at -80°C (up to 2 years).
  • Potent aldose reductase inhibition (IC50 = 35 nM).
  • Suitable as an analytical standard for in vitro assays.
  • Good solubility in DMSO and formulatable for aqueous use.
  • Long-term stability when stored under recommended conditions.
  • Defined chemical identity and molecular properties for reproducible results.
Catalog No. 50-002-89284