Learn More
Medchemexpress LLC N-(8-methoxy-1,2,3,4-tetrahydronaphthalen-2-yl)propanamide | 134865-70-6 | 99.3% | 233.31 g/mol | C14H19NO2 | 1 ML

Supplier: Medchemexpress LLC HY10135810MM 1ML
8-M-PDOT (N-(8-methoxy-1,2,3,4-tetrahydronaphthalen-2-yl)propanamide) is a selective melatonin MT2 receptor agonist provided in both solid and DMSO solution formats for laboratory research. The solution format is commonly supplied as 10 mM in DMSO (1 mL). Reported purity is ~99.33% and molecular weight is 233.31 g/mol. Intended for research use only.
- Selective MT2 receptor agonist for receptor pharmacology studies.
- Available as a 10 mM solution in DMSO and as solid powders.
- High purity (~99.3%) suitable for analytical and biological experiments.
- Storage recommendations provided for both powder and in-solution formats.
- Solvent and formulation guidance included for in vitro and in vivo use.
- Molecular weight 233.31 g/mol for dosing and preparation calculations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.