Learn More
Medchemexpress LLC N-(9-fluorenylmethoxycarbonyl)-D-glutamic acid | 104091-09-0 | MFCD09037362 | 98.0% | 369.37 | C20H19NO6 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW022255100G
Encompass_Preferred
Fmoc-D-Glu-OH is the Fmoc-protected D-enantiomer of glutamic acid supplied as a powder for research use. It is used as a building block in peptide synthesis and can also be employed to prepare supramolecular hydrogels; typical purity is about 98%.
  • Fmoc-protected D-glutamic acid suitable for peptide synthesis.
  • Powder form for easy weighing and handling.
  • High purity (~98%) for reliable experimental results.
  • Useful for preparing supramolecular hydrogels and materials research.
  • Available in larger package sizes for scale-up and bulk applications.
Catalog No. 50-002-99615