Learn More
Medchemexpress LLC N-(9-fluorenylmethoxycarbonyl)-L-leucine | 35661-60-0 | MFCD00037133 | 99.9% | 353.42 g/mol | C21H23NO4 | 50 G
SDP
Supplier:  Medchemexpress LLC HY10106450G
Encompass_Preferred
Fmoc-leucine is the Fmoc-protected derivative of L-leucine used as a building block in solid-phase peptide synthesis and other organic syntheses. It is supplied as a high-purity amino acid derivative suitable for coupling reactions, analytical work, and method development.
  • High purity suitable for peptide synthesis and analytical applications.
  • Fmoc protection enables standard Fmoc deprotection and coupling cycles.
  • Stable solid form for accurate weighing and storage.
  • Available in multiple pack sizes to support small-scale and bulk preparations.
  • Well characterized with CAS 35661-60-0 and defined molecular weight.
Catalog No. 50-002-11180