Learn More
Medchemexpress LLC N-(9-fluorenylmethoxycarbonyl)-L-leucine-5,5,5-d3 | 538372-74-6 | MFCD03428197 | 99.3% | 356.43 | C21H20D3NO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101064S25MG
Encompass_Preferred
Fmoc-leucine-d3 is the N-(9-fluorenylmethoxycarbonyl) protected, deuterium-labeled form of L-leucine (5,5,5-d3). It is provided as a stable isotope-labeled amino acid for use in peptide synthesis, quantitative mass spectrometry, isotopic tracing, and analytical method development.
  • Deuterium-labeled for mass spectrometry differentiation and quantitation.
  • Fmoc-protected for use in solid-phase peptide synthesis workflows.
  • High chemical purity for reliable analytical and synthetic results.
  • Available in small milligram packs suitable for research and method development.
  • Functions as an internal standard and isotopic tracer in quantitative assays.
Catalog No. 50-002-63792