Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-allylglycine | 222725-35-1 | 97.0% | 337.38 g/mol | C20H19NO4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0363525G
Encompass_Preferred
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-allylglycine is an Fmoc-protected N-allylglycine derivative used as a protected amino acid intermediate in peptide synthesis and related organic chemistry research. It provides an Fmoc-protected amino group with an allyl substituent that enables orthogonal protection/deprotection strategies, facilitating selective modification during multi-step syntheses. The compound is identified by CAS 222725-35-1 and has formula C20H19NO4 (MW 337.38 g/mol). For research use only.
  • Fmoc-protected amino acid suitable for peptide coupling.
  • Allyl substituent enables orthogonal deprotection pathways.
  • Compatible with solid-phase and solution-phase synthesis workflows.
  • Well characterized by CAS 222725-35-1 and molecular weight 337.38 g/mol.
  • Intended for research use only; not for diagnostic or therapeutic use.
Catalog No. 50-003-01875