Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-benzyl-L-alanine | 672917-68-9 | MFCD30595178 | 99.6% | 401.45 g/mol | C25H23NO4 | 50 MG

Supplier: Medchemexpress LLC HYW07273250MG
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-benzyl-L-alanine is an Fmoc-protected N-benzyl derivative of L-alanine used as a building block in peptide synthesis and related research applications. It is supplied as a high-purity solid for research use.
- Fmoc-protected amino acid derivative.
- High purity (99.6%).
- Molecular weight 401.45 g/mol.
- Chemical formula C25H23NO4.
- Off-white to light yellow solid form.
- Recommended storage: powder -20°C (3 years), 4°C (2 years).
- Available in small laboratory pack sizes (25 MG, 50 MG, 100 MG).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.