Learn More
Medchemexpress LLC N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-methyl-D-leucine | 103478-63-3 | HY-W011088 | 99.5% | 367.44 | 10 G

Supplier: Medchemexpress LLC HYW01108810G
N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-methyl-D-leucine is a leucine derivative. Amino acids and amino acid derivatives, such as this product, have been used as ergogenic supplements and are recognized for their benefits as dietary substances. They can influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stressful tasks, and help prevent exercise-induced muscle damage.
- Leucine derivative
- Solid, white to off-white appearance
- Molecular formula C22H25NO4
- Molecular weight 367.44
- Melting point 108-116 °C
- Boiling point 537.3 °C at 760 mmHg
- Flash point 278.7 °C
- Store powder at -20 °C for 3 years or 4 °C for 2 years
- Store in solvent at -80 °C for 6 months or -20 °C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.