Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-methyl-L-threonine | 252049-06-2 | 99.9% | 355.38 g·mol⁻¹ | C20H21NO5 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW036329
Encompass_Preferred
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-methyl-L-threonine is an Fmoc-protected, N-methylated threonine derivative used as an amino-acid building block in peptide synthesis. It provides a protected amine for standard solid-phase peptide synthesis and introduces N-methylation to modulate peptide backbone conformation for structure-activity studies.
  • Fmoc-protected amine compatible with standard Fmoc SPPS protocols.
  • N-methylation introduces conformational constraint useful for SAR and stability studies.
  • Suitable for incorporation into synthetic peptides and peptidomimetics.
  • Characterized by molecular formula C20H21NO5 and molecular weight 355.38 g·mol⁻¹.
Catalog No. 50-003-02900