Learn More
Medchemexpress LLC N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-methyl-L-threonine | 252049-06-2 | MFCD02094575 | 99.9% | 355.38 g/mol | C20H21NO5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03632910G
Encompass_Preferred
N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-methyl-L-threonine is an Fmoc-protected, N-methylated threonine derivative used as a protected amino acid building block in solid-phase peptide synthesis and related amino-acid research. It is suitable for gram-scale research applications and is reported with high purity and defined molecular properties.
Catalog No. 50-002-99459