Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-O-trityl-L-homoserine | 111061-55-3 | MFCD00270544 | 583.67 g/mol | C38H33NO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW048668100MG
Encompass_Preferred
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-O-trityl-L-homoserine is an Fmoc-protected O-trityl homoserine derivative used as a building block in peptide synthesis and related organic synthesis. It is supplied as a powder with molecular formula C38H33NO5 and molecular weight 583.67 g/mol.
  • Protected amino acid for peptide synthesis
  • Fmoc and trityl protecting groups enable selective deprotection
  • Powder form suitable for long-term storage at -20°C
  • Serves as a serine/homoserine derivative building block
  • CAS number 111061-55-3 for unambiguous identification
Catalog No. 50-002-10875