Learn More
Medchemexpress LLC N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-S-(4-(tert-butoxy)-4-oxobutyl)-L-cysteine | 102971-73-3 | MFCD11226818 | 98.7% | 485.59 g/mol | C26H31NO6S | 250 MG

Supplier: Medchemexpress LLC 250MGHYW048681
N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-S-(4-(tert-butoxy)-4-oxobutyl)-L-cysteine is an Fmoc-protected L-cysteine derivative used as a protected amino acid building block for peptide synthesis and related research applications. It is supplied as a high-purity powder with defined molecular properties for laboratory use.
- Fmoc-protected amino acid suitable for peptide synthesis.
- High purity (98.7%) for research applications.
- Molecular formula C26H31NO6S and molecular weight 485.59 g/mol.
- Available in multiple sizes, including 250 mg, 100 mg, 1 g, 5 g, and 10 g.
- Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (6 months) or -20°C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.