Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-S-trityl-L-homocysteine | 167015-23-8 | 99.8% | 599.74 g/mol | C38H33NO4S | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0489185G
Encompass_Preferred
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-S-trityl-L-homocysteine is an Fmoc- and S-trityl-protected derivative of L-homocysteine used as a building block in peptide and small-molecule synthesis. It provides an Fmoc-protected amino group and an S-trityl-protected thiol to allow orthogonal deprotection strategies during multi-step syntheses.
  • Fmoc- and S-trityl-protected homocysteine suitable for solid-phase peptide synthesis.
  • High reported purity (99.78%) supports consistent synthetic performance.
  • Available in multiple package sizes for small-scale synthesis and scale-up.
  • Documented CAS number 167015-23-8, molecular weight 599.74 g/mol, formula C38H33NO4S.
  • Datasheet, COA, and SDS available for handling and quality information.
Catalog No. 50-002-10732