Learn More
Medchemexpress LLC N-[(9H-fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine | 122889-11-6 | MFCD00237032 | 99.9% | 417.45 g/mol | C25H23NO5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02222610G
Encompass_Preferred
N-[(9H-fluoren-9-ylmethoxy)carbonyl]-O-(phenylmethyl)-D-serine is a protected serine derivative supplied as a solid reagent for research use. It contains an N-Fmoc protecting group and an O-benzyl (phenylmethyl) side-chain protection, making it suitable as a building block in peptide synthesis and related organic chemistry workflows.
  • Protected serine derivative suitable for peptide synthesis.
  • Contains N-Fmoc and O-benzyl protections for orthogonal deprotection.
  • High purity, reported as 99.89%.
  • Available in multiple package sizes for scale flexibility.
  • Provided for research use only.
Catalog No. 50-003-01241