Learn More
Medchemexpress LLC N-(9H-fluoren-9-ylmethoxycarbonyl)-L-allothreonine | 146306-75-4 | 99.1% | 341.36 g/mol | C19H19NO5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0486805G
Encompass_Preferred
N-(9H-fluoren-9-ylmethoxycarbonyl)-L-allothreonine is an Fmoc-protected allo-threonine derivative used as a protected amino acid building block in solid-phase peptide synthesis and related peptide chemistry.
  • Fmoc-protected amino acid for solid-phase peptide synthesis.
  • Suitable for manual and automated peptide assembly.
  • Typical purity near 99% (varies by batch).
  • Available in small laboratory pack sizes for research use.
  • Stored at room temperature unless otherwise specified in supplier documentation.
Catalog No. 50-002-10317