Learn More
Medchemexpress LLC N-acetyl-2-aminophenol | 614-80-2 | MFCD00002181 | 99.6% | 151.16 g/mol | C8H9NO2 | 50 G

Supplier: Medchemexpress LLC HYW01560050G
2-Acetamidophenol (N-acetyl-2-aminophenol) is a small-molecule organic compound supplied as an off-white to light-brown solid for research use. It has CAS 614-80-2, molecular formula C8H9NO2, and is offered in solid pack sizes such as 50 g. The material is provided at high purity (99.61%) and is suitable for biochemical and small-molecule studies.
- High-purity solid suitable for biochemical research.
- Molecular formula C8H9NO2 and molecular weight 151.16 g/mol.
- Commonly used as a reagent or intermediate in small-molecule studies.
- Supplied in multiple solid pack sizes including 50 g.
- Soluble in DMSO and available as ready-to-use DMSO solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.