Learn More
Medchemexpress LLC N-acetyl-2'-deoxy-2'-fluorocytidine | 159414-97-8 | MFCD15145138 | 99.6% | 287.24 g/mol | C11H14FN3O5 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1317531G
Encompass_Preferred
2'-F-Ac-C (N-acetyl-2'-deoxy-2'-fluorocytidine) is a fluorinated deoxycytidine nucleoside analog used as a building block in the synthesis of oligoribonucleotides and for research involving modified nucleic acids. The compound has a molecular weight of 287.24 g/mol and is supplied with analytical documentation supporting its quality.
  • Nucleoside analog suitable for oligoribonucleotide synthesis.
  • High chemical purity (99.6% by HPLC), supported by COA data.
  • Fluorinated deoxycytidine derivative for incorporation into modified oligonucleotides.
  • Storage stability when kept at -20°C under inert atmosphere; in solvent: -80°C up to 6 months.
  • Available in multiple pack sizes including gram-scale for synthetic applications.
Catalog No. 50-002-83266