Learn More
Medchemexpress LLC N-acetyl-β-alanine | 3025-95-4 | 98.0% | C5H9NO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY6600710G
Encompass_Preferred
N-Acetyl-β-alanine is identified as an abnormal amino acid metabolite and functions as a mono-N-protected amino acid (MPAA) ligand. This compound is also recognized as a human endogenous metabolite.
  • Abnormal amino acid metabolite
  • Functions as a mono-N-protected amino acid (MPAA) ligand
  • Recognized as a human endogenous metabolite
  • Purity of 98.0%
Catalog No. 50-004-34245