Learn More
CPC Scientific Ac-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-NH2 (ammonium salt) 1MG
SDP
Supplier:  CPC Scientific CCKS005B
Encompass
SEQUENCE: Ac-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-NH2 (ammonium salt)ONE-LETTER SEQUENCE: Ac-D-Tyr(SO3H)-MGWM-NH2MOLECULAR FORMULA: C38H50N8O13S3MOLECULAR WEIGHT: 923.1STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [89911-65-9]SYNONYMS: RESEARCH AREA: GastrointestinalREFERENCES:SKU(s): CCKS-005A, CCKS-005B
Catalog No. 50-210-9118