Learn More
Medchemexpress LLC N-Acetyl-D-glucosamine-13C8,15N | 95.0% | 13C8H1515NO6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYA0132S131MG
Encompass_Preferred
N-Acetyl-D-glucosamine-13C8,15N is a 13C and 15N labeled version of N-Acetyl-D-glucosamine, a monosaccharide derivative of glucose. It is intended for research use only. Stable heavy isotopes like those in this product are incorporated into drug molecules for quantitation during drug development, and deuteration, for example, can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-32789