Learn More
AdipoGen N-Acetyl-D-glucosamine (5 g)
SDP
Supplier:  AdipoGen AGCN20489G005
Encompass_Preferred
Chemical. CAS: 7512-17-6. Formula: C8H15NO6. MW: 221.2. Prepared by using chitin as a substrate. N-Acetyl-D-glucosamine (D-GlcNAc) is a non-toxic derivatized glucose monosaccharide found in polymers (Peptidoglycan; PGN) of bacterial cell walls, chitin (e.g. fungi, invertebrates, crustaceans), hyaluronic acids and various glycans. It is also synthesized in the glycosylation pathway as uridine diphosphate (UDP-GlcNAc), which can then be released following degradation of glycosylated proteins. Atypical microbial danger signal that acts as a new activator of NLRP3 inflammasome by dissociating the enzyme hexokinase from the mitochondria. D-GlcNAc inhibits purified hexokinase, which is also involved in the glucose metabolism and obesity in mM range. For hexokinase dissociation from the mitochondria much higher concentrations are needed.
Catalog No. 50-205-3673