Learn More
Medchemexpress LLC N-acetyl-D-glutamic acid | 19146-55-5 | MFCD00063196 | 97.0% | 189.17 g/mol | C7H11NO5 | 10 G

Supplier: Medchemexpress LLC HYW01524110G
N-acetyl-D-glutamic acid is the N-acetyl derivative of D-glutamic acid supplied as a solid research reagent for biochemical and peptide chemistry applications. It is intended for laboratory research use and is accompanied by storage recommendations and batch documentation for analytical confirmation.
- N-acetyl derivative of D-glutamic acid.
- Solid form for ease of handling and storage.
- Molecular weight 189.17 g/mol.
- Chemical formula C7H11NO5.
- Storage recommendations provided to maintain stability.
- Suitable for peptide synthesis and biochemical research applications.
- Batch-specific analytical data available on request.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.