Learn More
Gold Biotechnology Inc N-Acetyl-D-mannosamine 100 g
SDP
Supplier:  Gold Biotechnology Inc A152100
Encompass
N-Acetyl-D-mannosamine (ManNAc) is a specific substrate for the synthesis of N-acetylneuraminic acid, the essential precursor of bacterial capsular polysialic acid (PA). N-Acetyl-D-mannosamine is used for the synthesis of sialic acid. It is also a synthetic intermediate for a number of carbohydrate-derived families of biologically active compounds and pharmaceutical candidates. It is studied for its effect on capsular polysialic acid production.
Catalog No. NC1712421