Learn More
Selleck Chemical LLC N-Acetyl-D-mannosamine S3371-25mg
SDP
Supplier:  Selleck Chemical LLC S337125MG
Encompass_Preferred
N-Acetyl-D-mannosamine (ManNAc N-acetyl-beta-mannosamine) a precursor of a sialic acid improves the age-dependent attenuation of synaptic transmission and long-term potentiation (LTP) N-Acetyl-D-mannosamine treatment is a potential therapeutic application for cognitive dysfunction
Catalog No. 50-311-0778