Learn More
Medchemexpress LLC N-acetyl-D-proline | 59785-68-1 | ≥97.0% | 157.17 g/mol | C7H11NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0172585G
Encompass_Preferred
N-Acetyl-D-proline is the N-acetylated D-isomer of the amino acid proline supplied for biochemical and research use. It is a small molecule reagent (C7H11NO3, MW 157.17 g/mol) used in peptide chemistry, analytical reference applications, and general biochemical assays. Intended for laboratory research use.
  • N-acetylated D-isomer of proline.
  • Used as a reagent in peptide synthesis and modification.
  • Serves as an analytical reference or standard in assays.
  • Provided as a solid suitable for laboratory handling and storage.
  • Intended for research use only.
Catalog No. 50-003-00576