Learn More
Medchemexpress LLC N-acetyl-D-proline | 59785-68-1 | MFCD00080841 | 98.0% | 157.17 g/mol | C7H11NO3 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW017258100G
Encompass_Preferred
N-acetyl-D-proline is the N-acetyl derivative of D-proline supplied as a biochemical reagent for research and synthesis. It is a white to off-white solid commonly used as an intermediate in organic synthesis, peptide chemistry, and biochemical investigations.
  • High purity suitable for research use.
  • Molecular formula C7H11NO3; molecular weight 157.17 g/mol.
  • White to off-white solid, stable at room temperature.
  • Suitable as an intermediate in organic synthesis and peptide chemistry.
  • Available in laboratory-scale pack sizes for bench use.
Catalog No. 50-003-01186