Learn More
Cayman Chemical N-Acetyl-DL-AspartIc AcId 25g
SDP
Supplier:  Cayman Chemical 3463525 G
Encompass_Preferred
An acetylated form of L-aspartic acid; levels are increased in the brain of patients with Canavan disease
Catalog No. 50-294-7393