Learn More
Medchemexpress LLC N-acetyl-DL-methionine | 1115-47-5 | MFCD00008681 | 191.25 | C7H13NO3S | 10 G

Supplier: Medchemexpress LLC HYW01970410G
N-acetyl-DL-methionine is the N-acetylated derivative of the amino acid methionine, supplied for research use as an endogenous metabolite standard. The material is provided as a powder suitable for biochemical and analytical applications, with recommended storage conditions to preserve stability.
- Provided as a stable powder for storage and handling.
- Suitable as an endogenous metabolite standard for biochemical and analytical workflows.
- Soluble in common solvents for preparation of stock solutions.
- Recommended storage: powder at -20°C for long-term, 4°C for short-term; in solvent at -80°C for extended storage.
- Available in multiple pack sizes, including a 10 g option.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.