Learn More
Medchemexpress LLC N-acetyl-DL-serine | 97-14-3 | MFCD00037248 | 98.1% | 147.13 g·mol⁻1 | C5H9NO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW14209210MG
Encompass_Preferred
N-acetyl-DL-serine is an N-acetylated derivative of the amino acid serine supplied as a high-purity research reagent. It has reported antimicrobial activity and is used in biochemical and microbiological studies; it is provided as a water-soluble solid for preparation of stock solutions.
  • High purity (98.1%) suitable for research applications.
  • Water-soluble (125 mg/mL) for easy solution preparation.
  • Well-characterized chemical with CAS 97-14-3 for unambiguous identification.
  • Suitable for antimicrobial assays and biochemical studies.
  • Available in small, accurately measured packs for precise dosing.
Catalog No. 50-002-83690