Learn More
Medchemexpress LLC N-acetyl-DL-serine | 97-14-3 | MFCD00037248 | 98.1% | 147.13 g/mol | C5H9NO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW14209250MG
Encompass_Preferred
N-Acetyl-DL-serine is an N-acetylated amino acid supplied for research use, with reported antimicrobial properties and applications in molecular biology and biochemistry. It is provided as a small research quantity suitable for laboratory experiments.
  • High purity (98.1%) suitable for research.
  • Antimicrobial activity against Bacillus cereus and Staphylococcus aureus.
  • Useful for immobilizing DNA fragments on solid surfaces.
  • Applicable in protein synthesis studies and optical detection of DNA strands.
  • Good water solubility (125 mg/mL); may require ultrasonic assistance.
  • Molecular formula C5H9NO4; molecular weight 147.13 g/mol.
  • Supplied as a 50 mg research-grade sample.
Catalog No. 50-002-82306