Learn More
Medchemexpress LLC N-Acetyl-Ile-Glu-Pro-Asp-p-nitroanilide | 216757-29-8 | 99.8% | 634.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP32365MG
Encompass_Preferred
N-Acetyl-Ile-Glu-Pro-Asp-p-nitroanilide (Ac-IEPD-pNA) is a colorimetric peptide substrate of granzyme B designed for the accurate measurement of granzyme B activity.
  • Colorimetric peptide substrate
  • Accurately measures granzyme B activity
  • Solid appearance, white to off-white
  • For research use only
Catalog No. 50-003-97223