Learn More
Medchemexpress LLC N-acetyl-L-arginine | 155-84-0 | MFCD00019733 | 100.0% | 216.24 g·mol⁻¹ | C8H16N4O3 | 500 MG

Supplier: Medchemexpress LLC HYW014130500MG
N-acetyl-L-arginine is an acetylated derivative of the amino acid L-arginine used as a research reagent in biochemical and pharmacological studies. It is supplied for laboratory assays involving arginine metabolism, enzymatic activity, and studies of protein aggregation, and is available in powder and solution formats for experimental flexibility.
- Research-grade endogenous metabolite for biochemical studies.
- High purity: 99.97%.
- Molecular formula: C8H16N4O3.
- Molecular weight: 216.24 g·mol⁻¹.
- Available in multiple pack sizes, including 500 mg and 1 g.
- Supplied as powder or solution formats for assay flexibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.