Learn More
Medchemexpress LLC N-acetyl-L-aspartic acid, 4-(1,1-dimethylethyl) ester | 117833-18-8 | MFCD00236746 | 99.5% | 231.25 g/mol | C10H17NO5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02455410G
Encompass_Preferred
AC-Asp(OtBU)-OH is an N-acetylated aspartic acid derivative (N-acetyl-L-aspartic acid, tert-butyl ester) used as an amino-acid building block in peptide and small-molecule synthesis. Supplied as a high-purity solid, it is intended for research applications requiring a protected aspartic acid residue.
  • Provides N-acetylated aspartic acid building block for peptide synthesis.
  • Protects side-chain carboxyl group as a tert-butyl ester during coupling.
  • Offers high purity for reliable coupling and analytical results.
  • Available in multiple sizes for small-scale or bulk synthesis.
  • Stable under recommended storage conditions to preserve integrity.
Catalog No. 50-003-01623