Learn More
Medchemexpress LLC N-Acetyl-L-aspartic acid | 997-55-7 | 98.07% | 175.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113524R100MG
Encompass_Preferred
N-Acetyl-L-aspartic acid is an endogenous compound and a derivative of Aspartic acid. It functions as an acetyl donor and is involved in brain metabolism. This compound is utilized in research concerning neurodegenerative diseases, such as Canavan disease. It is intended for research and analytical applications.
  • Endogenous compound
  • Derivative of aspartic acid
  • Functions as an acetyl donor
  • Involved in brain metabolism
  • Utilized in research concerning neurodegenerative diseases, such as Canavan disease
Catalog No. 50-004-38466