Learn More
Medchemexpress LLC N-Acetyl-L-aspartic acid | 997-55-7 | 99.6% | 175.14 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY11352410MM/1ML
Encompass_Preferred
N-Acetyl-L-aspartic acid is a derivative of aspartic acid and an endogenous compound. It acts as an acetyl donor and is involved in brain metabolism. It is used in the research of neurodegenerative diseases such as Canavan disease.
  • Derivative of aspartic acid
  • Endogenous compound
  • Acts as an acetyl donor
  • Involved in brain metabolism
  • Used in the research of neurodegenerative diseases such as Canavan disease
Catalog No. 50-004-36386