Learn More
Medchemexpress LLC N-acetyl-L-aspartic acid | 997-55-7 | 99.6% | 175.14 | 50MG
SDP
Supplier:  Medchemexpress LLC HY113524R50MG
Encompass_Preferred
N-Acetyl-L-aspartic acid is a derivative of Aspartic acid and an endogenous compound involved in brain metabolism. It acts as an acetyl donor and is utilized in the research of neurodegenerative diseases, including Canavan disease.
  • Derivative of Aspartic acid
  • Acts as an acetyl donor
  • Involved in brain metabolism
  • Used in neurodegenerative disease research
  • Appears as a white to off-white solid
  • Soluble in water and DMSO
Catalog No. 50-004-36384