Learn More
Medchemexpress LLC N-Acetyl-L-aspartic acid-d3 | 284665-15-2 | 99.8% | 178.16 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY113524S1MG
Encompass_Preferred
N-Acetyl-L-aspartic acid-d3 is the deuterium labeled N-Acetyl-L-aspartic acid. It is a derivative of Aspartic acid and an endogenous compound. It acts as an acetyl donor and is involved in brain metabolism. N-Acetyl-L-aspartic acid is used in the research of neurodegenerative diseases such as Canavan disease.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36515