Learn More
Medchemexpress LLC N-Acetyl-L-aspartic acid (Standard) | 997-55-7 | >98.1% | 175.14 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY113524500MG
Encompass_Preferred
N-Acetyl-L-aspartic acid is a derivative of Aspartic acid and an endogenous compound crucial for brain metabolism, acting as an acetyl donor. It is extensively utilized in the research of neurodegenerative diseases, particularly Canavan disease.
  • Acts as an acetyl donor in brain metabolism
  • Used in research for neurodegenerative diseases
  • Demonstrated no adverse toxicological, neurobehavioral, or histopathological changes in 90-day rat studies
  • Did not cause adverse reproductive, developmental, or ophthalmological effects in rat studies
Catalog No. 50-004-36438