Learn More
Medchemexpress LLC N-Acetyl-L-aspartic acid (Standard) | 997-55-7 | 99.6% | 175.14 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY113524R10MG
Encompass_Preferred
N-Acetyl-L-aspartic acid (Standard) is an analytical standard intended for research and analytical applications. It acts as an acetyl donor, is involved in brain metabolism, and is used in the research of neurodegenerative diseases, such as Canavan disease.
  • Derivative of aspartic acid
  • Endogenous compound
  • Acts as an acetyl donor
  • Involved in brain metabolism
  • Used in research of neurodegenerative diseases
  • Analytical standard for assays
  • Commonly used in HPLC, GC, and MS experiments
  • Appears as a solid, white to off-white in color
Catalog No. 50-004-37470