Learn More
Medchemexpress LLC N-acetyl-L-cysteine ethyl ester | 59587-09-6 | MFCD09954466 | 99.9% | 191.25 g/mol | C7H13NO3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1344955MG
Encompass_Preferred
N-Acetyl-L-cysteine ethyl ester (NACET) is the ethyl-ester derivative of N-acetyl-L-cysteine with enhanced membrane permeability. It is provided as a research-grade small molecule solid for biochemical and cell-based studies involving thiol delivery, glutathione metabolism, and modulation of reactive oxygen species.
  • Improved cell permeability compared with the parent compound.
  • Precursor to N-acetylcysteine and intracellular glutathione.
  • Useful in oxidative stress and redox biology studies.
  • Suitable for small-scale exploratory experiments and assays.
  • Supplied as a solid with defined molecular weight and chemical formula.
Catalog No. 50-002-22024