Learn More
Alkali Scientific n Acetyl L cysteine (NAC), 500 G
SDP
Supplier:  Alkali Scientific CET003CP
Encompass_Preferred
Acetyl L-cysteine (NAC) is a powerful antioxidant that helps maintain cell viability by preventing oxidative damage and supporting the synthesis of glutathione, a key cellular protectant. This 500g package is designed for larger-scale cell culture and research applications. NAC is used in studies involving oxidative stress, detoxification, and cellular damage prevention. It is commonly used in biochemical research, drug discovery, and cellular biology to protect cells during exposure to toxins, environmental stress, and in drug testing protocols. The 500g size ensures a sufficient supply for extended experiments and high-volume research needs.
Catalog No. 50-006-60343