Learn More
Medchemexpress LLC HY-W015240 500mg Medchemexpress, N-Acetyl-L-glutamic acid CAS:1188-37-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW015240500MG
Encompass_Preferred
Medchemexpress, HY-W015240 500mg N-Acetyl-L-glutamic acid CAS:1188-37-0 N-Acetyl-L-glutamic acid, a N-acyl-L-amino acid, is a component of animal cell culturing media. N-Acetyl-L-glutamic acid is a metabolite of Saccharomyces cerevisiae and human. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3371