Learn More
Cayman Chemical N-acetyl-L-IsoleucIn 250g
SDP
Supplier:  Cayman Chemical 35545250 G
Encompass_Preferred
An acetylated form of L-isoleucine; urine levels are increased in patients with MSUD
Catalog No. 50-291-0088