Learn More
Cayman Chemical N-acetyl-L-OrnIthIn 50mg
SDP
Supplier:  Cayman Chemical 3609250 MG
Encompass_Preferred
An intermediate in arginine and proline metabolism; a precursor in the synthesis of γ-glutamic semialdehyde
Catalog No. 50-291-3198