Learn More
Medchemexpress LLC N-Acetyl-L-proline | 68-95-1 | 157.17 | 10 G

Supplier: Medchemexpress LLC HYW00411410G
N-Acetyl-L-proline is a proline derivative and human endogenous metabolite. It is for research use only and not sold to patients.
- Purity: 99.92%
- Molecular weight: 157.17
- Formula: C7H11NO3
- CAS number: 68-95-1
- Appearance: Solid
- Color: White to off-white
- Structure classification: Alkaloids, pyrrole alkaloids
- Initial source: Endogenous metabolite
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year
- Solubility (in vitro): DMSO: ≥ 100 mg/mL (636.25 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.