Learn More
Medchemexpress LLC N-Acetyl-L-proline | 68-95-1 | 157.17 | 100 G

Supplier: Medchemexpress LLC HYW004114100G
N-Acetyl-L-proline is a proline derivative and an endogenous metabolite. It is intended for research use only and not for patient use.
- Purity: 99.92%
- Molecular weight: 157.17
- Formula: C7H11NO3
- CAS no.: 68-95-1
- Appearance: Solid
- Color: White to off-white
- Structure classification: Alkaloids, pyrrole alkaloids
- Initial source: Endogenous metabolite
- Shipping at room temperature in continental US; may vary elsewhere
- Solid storage: Store at room temperature for 3 years
- In solvent storage: -80°C for 2 years or -20°C for 1 year
- Soluble in DMSO at ≥ 100 mg/mL (636.25 mM)
- Used in research involving amino acid derivatives
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.